Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Iodononane, 97%, stab. with copper

Chemical Identifiers

Chemical Name or Material 1-Iodononane
CAS 4282-42-2
Name Note Stabilized with copper
Molecular Formula CH3(CH2)8I
Formula Weight 254.16
MDL Number MFCD00001107
Beilstein 1733634
Synonym n-Nonyl iodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1066714

 
alfa aesar™
A1066714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 254.16
Chemical Name or Material 1-Iodononane
Boiling Point 107° to 108°C (8mmHg)
TSCA TSCA
Name Note Stabilized with copper
Assay 97%
Molecular Formula CH3(CH2)8I
MDL Number MFCD00001107
CAS 4282-42-2
Quantity 25g
Beilstein 1733634
Synonym n-Nonyl iodide
Flash Point 85°C (185°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.