missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Iodoisoquinoline 98.0+%, TCI America™
$413.10 - $1,595.95
Chemical Identifiers
CAS | 19658-77-6 |
---|---|
Molecular Formula | C9H6IN |
Molecular Weight (g/mol) | 255.06 |
MDL Number | MFCD00234494 |
InChI Key | FDDBUIWRNBGXHB-UHFFFAOYSA-N |
PubChem CID | 640964 |
IUPAC Name | 1-iodoisoquinoline |
SMILES | IC1=NC=CC2=CC=CC=C12 |
Chemical Identifiers
19658-77-6 | |
255.06 | |
FDDBUIWRNBGXHB-UHFFFAOYSA-N | |
1-iodoisoquinoline |
C9H6IN | |
MFCD00234494 | |
640964 | |
IC1=NC=CC2=CC=CC=C12 |
Specifications
19658-77-6 | |
C9H6IN | |
5 g | |
IC1=NC=CC2=CC=CC=C12 | |
255.06 | |
255.06 | |
Crystalline Powder |
75°C | |
MFCD00234494 | |
FDDBUIWRNBGXHB-UHFFFAOYSA-N | |
1-iodoisoquinoline | |
640964 | |
≥98.0% (GC,T) | |
1-Iodoisoquinoline |
Safety and Handling
TSCA : No