Please login to your online account to display your discounted pricing

1-Iododecane, 98%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  33036-1000

 View more versions of this product

Catalog No. AC330361000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (GC)
Physical Form Liquid
Formula Weight 268.17
Quantity 100g
Color Clear colorless
Density 1.2500g/mL
CAS Max % 100.0
Linear Formula CH3(CH2)9I
Infrared Spectrum Authentic
CAS 2050-77-3
Molecular Formula C10H21I
Assay Percent Range 98%
MDL Number MFCD00001085
CAS Min % 97.5
Synonym Decyl iodide
Refractive Index 1.4840 to 1.4870 (20°C, 589nm)
Packaging Glass bottle
Chemical Name or Material 1-Iododecane
Stabilizer copper
Health Hazard 2 GHS P Statement Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation.
Health Hazard 3 GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection.

Specifications

Chemical Name or Material 1-Iododecane
CAS 2050-77-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C10H21I
Linear Formula CH3(CH2)9I
Formula Weight 268.17
MDL Number MFCD00001085
Synonym Decyl iodide
Accessories

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection.

GHS P Statement Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation.

SDS & Safety Documents