Please login to your online account to display your discounted pricing

1-Iodo-4-(trifluoromethyl)benzene, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  TL00447EA

 View more versions of this product

Catalog No. TL00447EA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 272
Quantity 10g
Assay 97%
Density 1.8510g/mL
CAS Min % 97.0
Synonym 1-Iodo-4-(trifluoromethyl)benzene
Packaging Amber Glass Bottle
Linear Formula IC6H4CF3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Molecular Formula C7H4F3I
MDL Number MFCD00039398
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Chemical Name or Material 1-Iodo-4-(trifluoromethyl)benzene
CAS 455-13-0

Specifications

Chemical Name or Material 1-Iodo-4-(trifluoromethyl)benzene
CAS 455-13-0
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C7H4F3I
Linear Formula IC6H4CF3
Formula Weight 272
MDL Number MFCD00039398
Synonym 1-Iodo-4-(trifluoromethyl)benzene
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents