Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Iodo-3-(trimethylsiloxy)benzene, 98%

$189.85 - $189.85

Chemical Identifiers

Chemical Name or Material 1-Iodo-3-(trimethylsiloxy)benzene
Molecular Formula C9H13IOSi
Formula Weight 292.19
MDL Number MFCD03788743
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1958803

  SDS
alfa aesar™
L1958803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1958806

  SDS
alfa aesar™
L1958806
5g Each for $189.85
Description & Specifications

Specifications

Formula Weight 292.19
MDL Number MFCD03788743
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Molecular Formula C9H13IOSi
Health Hazard 1 Warning
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Chemical Name or Material 1-Iodo-3-(trimethylsiloxy)benzene
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.