Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Iodo-3-(trifluoromethoxy)benzene, 97+%

$83.49 - $334.50

Chemical Identifiers

Chemical Name or Material 1-Iodo-3-(trifluoromethoxy)benzene
CAS 198206-33-6
Molecular Formula C7H4F3IO
Formula Weight 288.01
MDL Number MFCD01090992
Synonym 3-(Trifluoromethoxy)iodobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2416903

  SDS
alfa aesar™
B2416903
1g Each for $83.49

AAB2416906

  SDS
alfa aesar™
B2416906
5g Each for $334.50
Description & Specifications

Specifications

Formula Weight 288.01
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1-Iodo-3-(trifluoromethoxy)benzene
Assay ≥97%
Quantity 1g
Health Hazard 1 Warning
Synonym 3-(Trifluoromethoxy)iodobenzene
Flash Point 65°C (150°F)
Molecular Formula C7H4F3IO
Boiling Point 185° to 186°C
MDL Number MFCD01090992
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 198206-33-6
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.