missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Iodo-3-methylbutane, 97%, stab. with copper

Chemical Identifiers

CAS 541-28-6
Molecular Formula C5H11I
Formula Weight 198.05
MDL Number MFCD00039414
UN Number UN1993
Beilstein 1730990
Merck Index 14,5120
Synonym Isoamyl iodide; Isopentyl iodide
Chemical Name or Material 1-Iodo-3-methylbutane, stabilized with copper
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0519409 SDS Alfa Aesar™
L0519409
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAL0519418 SDS Alfa Aesar™
L0519418
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
1-Iodo-3-methylbutane, 97%, stab. with copper can be used for organic synthesis or used as solvents.

Solubility
Insoluble in water. Soluble in most common organic solvents.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Description & Specifications

Chemical Identifiers

541-28-6
198.05
UN1993
14,5120
1-Iodo-3-methylbutane, stabilized with copper
C5H11I
MFCD00039414
1730990
Isoamyl iodide; Isopentyl iodide

Specifications

146° to 148°C
10g
C5H11I
MFCD00039414
1730990
Isoamyl iodide; Isopentyl iodide
97%
541-28-6
40°C (104°F)
198.05
UN1993
14,5120
1-Iodo-3-methylbutane, stabilized with copper

Safety and Handling

GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

RTECSNumber : EK4412000

TSCA : TSCA