Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Iodo-3,5-dinitrobenzene, 99%

$527.97 - $527.97

Chemical Identifiers

Chemical Name or Material 1-Iodo-3,5-dinitrobenzene
CAS 6276-04-6
Molecular Formula C6H3IN2O4
Formula Weight 294.01
MDL Number MFCD00024330
Synonym 3,5-Dinitroiodobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2817014

  SDS
alfa aesar™
H2817014
25g Each for $527.97
Description & Specifications

Specifications

Formula Weight 294.01
MDL Number MFCD00024330
Chemical Name or Material 1-Iodo-3,5-dinitrobenzene
Assay 99%
Molecular Formula C6H3IN2O4
CAS 6276-04-6
Melting Point 99° to 103°C
Synonym 3,5-Dinitroiodobenzene
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H317-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P262-P305+P351+P338
Wear protective gloves/clothing.
Do not get in eyes, on skin, or on clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Applications
As cytochemical reagent. For kinetic-potentiometric study and analytical applications of micellar-catalysed reactions.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.

Safety and Handling

Warning

GHS P Statement
P280h-P262-P305+P351+P338
Wear protective gloves/clothing.
Do not get in eyes, on skin, or on clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H312-H332-H315-H319-H317-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.