missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Iodo-2-(trimethylsilyl)acetylene, 97%

Manufacturer:  Alfa Aesar™ H5337514

 View more versions of this product

Catalog No. AAH5337514



Description

Description

Applications
1-Iodo-2-(trimethylsilyl)acetylene may be used to investigate the reactivity of diamagnetic cobalt(I) complex, Tol-BDI((2-pp)2)Co [Tol-BDI((2-pp)2)H = 2-(4-tolyl)-1,3-bis(2-isopropylpyridyl)propenediimine]. It may be used for the synthesis of 1-(4-trimethylsilanylbuta-1,3-diynyl)pyrene, via Pd-coupling with 1-iodo-2-trimethylsilylacetylene.

Notes
Light sensitive. Incompatible with oxidizing agents.
Specifications

Specifications

1-Iodo-2-(trimethylsilyl)acetylene
C5H9ISi
MFCD00274201
1-iodo-2-trimethylsilyl acetylene, iodoethynyl trimethylsilane, silane, iodomethyl trimethyl, 2-iodoethynyl trimethylsilane, 2-iodoethynyl trimethyl silane, silane, iodoethynyl trimethyl, tbdmcs, acmc-20aoug, trimethyliodoethinylsilane, iodoethynyltrimethylsilane
C[Si](C)(C)C#CI
224.116
224.12
130°C
48°C (118°F)
18163-47-8
(CH3)3SiC≡CI
UN1993
HNIRHTRSZDSMOF-UHFFFAOYSA-N
2-iodoethynyl(trimethyl)silane
140341
97%
25g

Chemical Identifiers

18163-47-8
224.116
HNIRHTRSZDSMOF-UHFFFAOYSA-N
140341
C[Si](C)(C)C#CI
C5H9ISi
MFCD00274201
1-iodo-2-trimethylsilyl acetylene, iodoethynyl trimethylsilane, silane, iodomethyl trimethyl, 2-iodoethynyl trimethylsilane, 2-iodoethynyl trimethyl silane, silane, iodoethynyl trimethyl, tbdmcs, acmc-20aoug, trimethyliodoethinylsilane, iodoethynyltrimethylsilane
2-iodoethynyl(trimethyl)silane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H334-H361-H226-H332-H317
Toxic if swallowed.
Toxic in contact with skin.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Flammable liquid and vapor.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

SDS Documents