Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Iodo-2-(trifluoromethoxy)benzene, 98%

$279.69 - $279.69

Chemical Identifiers

Chemical Name or Material 1-Iodo-2-(trifluoromethoxy)benzene
CAS 175278-00-9
Molecular Formula C7H4F3IO
Formula Weight 288.01g/mol
MDL Number MFCD00042410
Beilstein 8762170
Synonym 2-(Trifluoromethoxy)iodobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1564514

 
alfa aesar™
L15645-14
25g Each for $279.69
Description & Specifications

Specifications

Boiling Point 164° to 165°C
Quantity 25g
Flash Point 65°C (149°F)
Chemical Name or Material 1-Iodo-2-(trifluoromethoxy)benzene
CAS 175278-00-9
Assay 98%
Molecular Formula C7H4F3IO
Formula Weight 288.01g/mol
MDL Number MFCD00042410
Beilstein 8762170
Synonym 2-(Trifluoromethoxy)iodobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.