missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Iodo-2-nitrobenzene, 97%

Manufacturer:  Alfa Aesar™ A1210022

 View more versions of this product

Catalog No. AAA1210022



Specifications

Specifications

1-Iodo-2-nitrobenzene
288°C to 289°C
100g
C6H4INO2
2044110
JXMZUNPWVXQADG-UHFFFAOYSA-N
1-iodo-2-nitrobenzene
69115
97%
>110°C (230°F)
48°C to 52°C
609-73-4
MFCD00007088
2-iodonitrobenzene, o-iodonitrobenzene, o-nitroiodobenzene, benzene, 1-iodo-2-nitro, 2-nitroiodobenzene, 2-iodo-1-nitrobenzene, 1-iodo-2-nitro-benzene, 2-iodo-nitrobenzene, 2-nitro-iodobenzene, benzene, iodonitro
C1=CC=C(C(=C1)[N+](=O)[O-])I
249.007
249.01

Chemical Identifiers

609-73-4
249.007
JXMZUNPWVXQADG-UHFFFAOYSA-N
69115
C1=CC=C(C(=C1)[N+](=O)[O-])I
C6H4INO2
MFCD00007088
2-iodonitrobenzene, o-iodonitrobenzene, o-nitroiodobenzene, benzene, 1-iodo-2-nitro, 2-nitroiodobenzene, 2-iodo-1-nitrobenzene, 1-iodo-2-nitro-benzene, 2-iodo-nitrobenzene, 2-nitro-iodobenzene, benzene, iodonitro
1-iodo-2-nitrobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

SDS Documents