Learn More
1-Iodo-2-methylpropane, 97%, stab. with copper, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1588314
Description
1-Iodo-2-methylpropane is used as solvents, analytical reagents and organic synthesis of raw materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Iodo-2-methylpropane is used as solvents, analytical reagents and organic synthesis of raw materials.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Light sensitive.
Specifications
1-Iodo-2-methylpropane | |
513-38-2 | |
1.599 | |
12°C (53°F) | |
1.496 | |
25 g | |
1730927 | |
14,5144 | |
Insoluble in water. | |
CC(C)CI | |
184.02 | |
184.02 |
stabilized with copper | |
-93°C | |
120°C to 121°C | |
C4H9I | |
MFCD00001084 | |
UN2391 | |
Light sensitive | |
isobutyl iodide, propane, 1-iodo-2-methyl, isobutyljodid, isobutyliodide, 2-methylpropyl iodide, primary isobutyl iodide, isobutyljodid czech, 1-jod-2-methylpropan, 1-jod-2-methylpropan czech, 1-iodo-2-methyl-propane | |
BTUGGGLMQBJCBN-UHFFFAOYSA-N | |
1-iodo-2-methylpropane | |
10556 | |
98% |
Chemical Identifiers
513-38-2 | |
184.02 | |
BTUGGGLMQBJCBN-UHFFFAOYSA-N | |
10556 | |
CC(C)CI |
C4H9I | |
MFCD00001084 | |
isobutyl iodide, propane, 1-iodo-2-methyl, isobutyljodid, isobutyliodide, 2-methylpropyl iodide, primary isobutyl iodide, isobutyljodid czech, 1-jod-2-methylpropan, 1-jod-2-methylpropan czech, 1-iodo-2-methyl-propane | |
1-iodo-2-methylpropane |
Safety and Handling
GHS H Statement
H225-H332-H315-H319-H335
Highly flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H332-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: IODOMETHYLPROPANES
EINECSNumber : 208-160-5
RTECSNumber : TZ4250000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only