Please login to your online account to display your discounted pricing

1-Iodo-2-methylpropane, 97%, stabilized, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC223745000

 View more versions of this product

Catalog No. AC223745000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 184.02
Quantity 500mL
Color Clear colorless to light orange
EINECS Number 208-160-5
TSCA TSCA
Density 1.5900g/mL
RTECS Number TZ4250000
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.4948 to 1.4968 (20°C, 589nm)
Stabilizer copper
Synonym Isobutyl iodide
Health Hazard 2 GHS P Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Harmful if inhaled.
Chemical Name or Material 1-Iodo-2-methylpropane
Infrared Spectrum Authentic
Name Note 97%
Assay Percent Range 97%
Molecular Formula C4H9I
Linear Formula (CH3)2CHCH2I
MDL Number MFCD00001084
CAS 513-38-2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 1-Iodo-2-methylpropane
Name Note 97%
CAS 513-38-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H9I
Linear Formula (CH3)2CHCH2I
Formula Weight 184.02
MDL Number MFCD00001084
Synonym Isobutyl iodide
Accessories

GHS Signal Word: Danger

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Harmful if inhaled.

SDS & Safety Documents