missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Iodo-2-methylbutane (stabilized with Copper chip), TCI America™
$216.95 - $641.69
Chemical Identifiers
CAS | 616-14-8 |
---|---|
Molecular Formula | C5H11I |
Molecular Weight (g/mol) | 198.047 |
MDL Number | MFCD01321177 |
InChI Key | RHBHXHXNWHTGSO-UHFFFAOYSA-N |
PubChem CID | 141479 |
IUPAC Name | 1-iodo-2-methylbutane |
SMILES | CCC(C)CI |
Chemical Identifiers
616-14-8 | |
198.047 | |
RHBHXHXNWHTGSO-UHFFFAOYSA-N | |
1-iodo-2-methylbutane |
C5H11I | |
MFCD01321177 | |
141479 | |
CCC(C)CI |
Specifications
148°C | |
25g | |
C5H11I | |
1993 | |
CCC(C)CI | |
198.047 | |
198.05 | |
≥95.0% (GC) |
Undesignated | |
616-14-8 | |
MFCD01321177 | |
RHBHXHXNWHTGSO-UHFFFAOYSA-N | |
1-iodo-2-methylbutane | |
141479 | |
Clear Liquid at 20°C | |
1-Iodo-2-methylbutane (stabilized with Copper chip) |
Safety and Handling
TSCA : No