Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Iodo-2-isopropylbenzene, 95%

$79.72 - $79.72

Chemical Identifiers

Chemical Name or Material 1-Iodo-2-isopropylbenzene
CAS 19099-54-8
Molecular Formula C9H11I
Formula Weight 246.09
MDL Number MFCD00013704
Beilstein 2351872
Synonym 2-Iodocumene; 2-Isopropyliodobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0153306

  SDS
alfa aesar™
L0153306
5g Each for $79.72
Description & Specifications

Specifications

Formula Weight 246.09
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Synonym 2-Iodocumene; 2-Isopropyliodobenzene
Chemical Name or Material 1-Iodo-2-isopropylbenzene
Assay 95%
MDL Number MFCD00013704
Molecular Formula C9H11I
Boiling Point 110°C (20mmHg)
Quantity 5g
CAS 19099-54-8
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Health Hazard 1 Warning
Odor Aromatic
Beilstein 2351872
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.