missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Indoleacetic acid, 95%, Thermo Scientific Chemicals
$160.32 - $369.90
Chemical Identifiers
CAS | 24297-59-4 |
---|---|
Molecular Formula | C10H9NO2 |
Molecular Weight (g/mol) | 175.187 |
MDL Number | MFCD00047262 |
InChI Key | WQJFIWXYPKYBTO-UHFFFAOYSA-N |
Synonym | 1h-indole-1-acetic acid, 2-1h-indol-1-yl acetic acid, indol-1-yl-acetic acid, indole-1-acetic acid, 1h-indol-1-ylacetic acid, indol-1-ylacetic acid, indole-n-acetic acid, 1-indoleacetic acid, pubchem20663, 1-indolyl acetic acid |
PubChem CID | 90448 |
ChEBI | CHEBI:72814 |
IUPAC Name | 2-indol-1-ylacetic acid |
SMILES | C1=CC=C2C(=C1)C=CN2CC(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH63935MD
|
Thermo Scientific Chemicals
H63935MD |
250 mg |
Each for $160.32
|
|
|||||
AAH6393503
|
Thermo Scientific Chemicals
H6393503 |
1 g |
Each for $369.90
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
24297-59-4 | |
175.187 | |
WQJFIWXYPKYBTO-UHFFFAOYSA-N | |
90448 | |
2-indol-1-ylacetic acid |
C10H9NO2 | |
MFCD00047262 | |
1h-indole-1-acetic acid, 2-1h-indol-1-yl acetic acid, indol-1-yl-acetic acid, indole-1-acetic acid, 1h-indol-1-ylacetic acid, indol-1-ylacetic acid, indole-n-acetic acid, 1-indoleacetic acid, pubchem20663, 1-indolyl acetic acid | |
CHEBI:72814 | |
C1=CC=C2C(=C1)C=CN2CC(=O)O |
Specifications
24297-59-4 | |
MFCD00047262 | |
142924 | |
WQJFIWXYPKYBTO-UHFFFAOYSA-N | |
2-indol-1-ylacetic acid | |
90448 | |
175.18 | |
Solid |
C10H9NO2 | |
250 mg | |
1h-indole-1-acetic acid, 2-1h-indol-1-yl acetic acid, indol-1-yl-acetic acid, indole-1-acetic acid, 1h-indol-1-ylacetic acid, indol-1-ylacetic acid, indole-n-acetic acid, 1-indoleacetic acid, pubchem20663, 1-indolyl acetic acid | |
C1=CC=C2C(=C1)C=CN2CC(=O)O | |
175.187 | |
CHEBI:72814 | |
95% | |
1-Indoleacetic acid |
Safety and Handling
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only