missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Indoleacetic acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6393503
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Indoleacetic acid | |
C10H9NO2 | |
1 g | |
1h-indole-1-acetic acid, 2-1h-indol-1-yl acetic acid, indol-1-yl-acetic acid, indole-1-acetic acid, 1h-indol-1-ylacetic acid, indol-1-ylacetic acid, indole-n-acetic acid, 1-indoleacetic acid, pubchem20663, 1-indolyl acetic acid | |
C1=CC=C2C(=C1)C=CN2CC(=O)O | |
175.187 | |
CHEBI:72814 | |
95% |
24297-59-4 | |
MFCD00047262 | |
142924 | |
WQJFIWXYPKYBTO-UHFFFAOYSA-N | |
2-indol-1-ylacetic acid | |
90448 | |
175.18 | |
Solid |
Chemical Identifiers
24297-59-4 | |
175.187 | |
WQJFIWXYPKYBTO-UHFFFAOYSA-N | |
90448 | |
2-indol-1-ylacetic acid |
C10H9NO2 | |
MFCD00047262 | |
1h-indole-1-acetic acid, 2-1h-indol-1-yl acetic acid, indol-1-yl-acetic acid, indole-1-acetic acid, 1h-indol-1-ylacetic acid, indol-1-ylacetic acid, indole-n-acetic acid, 1-indoleacetic acid, pubchem20663, 1-indolyl acetic acid | |
CHEBI:72814 | |
C1=CC=C2C(=C1)C=CN2CC(=O)O |
Safety and Handling
P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c
H302-H318
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only