missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Indanone, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 122111000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Indanone | |
38.0°C to 42.0°C | |
1.1000g/mL | |
>110°C | |
99% min. (GC) | |
C9H8O | |
07,36 | |
1-indanone, indan-1-one, 2,3-dihydro-1h-inden-1-one, indanone, alpha-indanone, alpha-hydrindone, 1h-inden-1-one, 2,3-dihydro, 1-indone, hydrindone, .alpha.-indanone | |
QNXSIUBBGPHDDE-UHFFFAOYSA-N | |
2,3-dihydroinden-1-one | |
6735 | |
132.16 | |
Crystalline Mass |
83-33-0 | |
Brown to Yellow | |
243.0°C to 245.0°C | |
Authentic | |
Glass bottle | |
100 g | |
1.1 | |
Solubility in water: 6.5g/L (20°C). Other solubilities: soluble in ethanol,ether and chloroform | |
C1CC(=O)C2=CC=CC=C21 | |
132.16 | |
CHEBI:17404 | |
99+% |
Chemical Identifiers
83-33-0 | |
132.16 | |
1-indanone, indan-1-one, 2,3-dihydro-1h-inden-1-one, indanone, alpha-indanone, alpha-hydrindone, 1h-inden-1-one, 2,3-dihydro, 1-indone, hydrindone, .alpha.-indanone | |
CHEBI:17404 | |
C1CC(=O)C2=CC=CC=C21 |
C9H8O | |
QNXSIUBBGPHDDE-UHFFFAOYSA-N | |
6735 | |
2,3-dihydroinden-1-one |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wash face,hands and any exposed skin thoroughly after handling.
Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 201-470-1
RUO – Research Use Only