Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Indanol, 98%

$55.88 - $321.72

Chemical Identifiers

Chemical Name or Material 1-Indanol
CAS 6351-10-6
Molecular Formula C9H10O
Formula Weight 134.18
MDL Number MFCD00003797
Beilstein 2042960
Synonym 1-Hydroxyindane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2453606

  SDS
alfa aesar™
B2453606
5g Each for $55.88

AAB2453614

  SDS
alfa aesar™
B2453614
25g Each for $190.86

AAB2453618

  SDS
alfa aesar™
B2453618
50g Each for $321.72
Description & Specifications

Specifications

Formula Weight 134.18
Chemical Name or Material 1-Indanol
Assay 98%
Molecular Formula C9H10O
MDL Number MFCD00003797
CAS 6351-10-6
Beilstein 2042960
Synonym 1-Hydroxyindane
Health Hazard 1 Warning
Boiling Point 128°C (12mmHg)
Melting Point 50° to 54°C
Flash Point 145°C (293°F)
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
1-Indanol derivative was found to be an efficient ligand for the palladium-catalyzed asymmetric Heck reaction. It is used as pharmaceutical intermediate.

Solubility
Soluble in water 10 g/L at 20°C, ethanol, benzene.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.