missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Imidazoleacetic Acid 98.0+%, TCI America™
$33.17 - $127.75
Chemical Identifiers
CAS | 22884-10-2 |
---|---|
Molecular Formula | C5H6N2O2 |
Molecular Weight (g/mol) | 126.115 |
MDL Number | MFCD01721800 |
InChI Key | QAFBDRSXXHEXGB-UHFFFAOYSA-N |
Synonym | imidazol-1-yl-acetic acid, 2-1h-imidazol-1-yl acetic acid, 1h-imidazole-1-acetic acid, 1h-imidazol-1-ylacetic acid, imidazole-1-acetic acid, 1-imidazolyl acetic acid, 1-imidazoleacetic acid, imidazol-1-ylacetic acid, imidazol-1-yl acetic acid, 1-imidazolylacetic acid |
PubChem CID | 656129 |
ChEBI | CHEBI:70801 |
IUPAC Name | 2-imidazol-1-ylacetic acid |
SMILES | C1=CN(C=N1)CC(=O)O |
Chemical Identifiers
22884-10-2 | |
126.115 | |
QAFBDRSXXHEXGB-UHFFFAOYSA-N | |
656129 | |
2-imidazol-1-ylacetic acid |
C5H6N2O2 | |
MFCD01721800 | |
imidazol-1-yl-acetic acid, 2-1h-imidazol-1-yl acetic acid, 1h-imidazole-1-acetic acid, 1h-imidazol-1-ylacetic acid, imidazole-1-acetic acid, 1-imidazolyl acetic acid, 1-imidazoleacetic acid, imidazol-1-ylacetic acid, imidazol-1-yl acetic acid, 1-imidazolylacetic acid | |
CHEBI:70801 | |
C1=CN(C=N1)CC(=O)O |
Specifications
22884-10-2 | |
White-Yellow | |
MFCD01721800 | |
imidazol-1-yl-acetic acid, 2-1h-imidazol-1-yl acetic acid, 1h-imidazole-1-acetic acid, 1h-imidazol-1-ylacetic acid, imidazole-1-acetic acid, 1-imidazolyl acetic acid, 1-imidazoleacetic acid, imidazol-1-ylacetic acid, imidazol-1-yl acetic acid, 1-imidazolylacetic acid | |
C1=CN(C=N1)CC(=O)O | |
126.115 | |
CHEBI:70801 | |
≥98.0% (GC,T) | |
1-Imidazoleacetic Acid |
269°C | |
C5H6N2O2 | |
5 g | |
QAFBDRSXXHEXGB-UHFFFAOYSA-N | |
2-imidazol-1-ylacetic acid | |
656129 | |
126.12 | |
Crystalline Powder |
Safety and Handling
TSCA : No