Alfa Aesar™ 1-Hydroxytetraphenylcyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldiruthenium(II), 98%
Manufacturer:  Alfa Aesar™ 4688404
 View more versions of this product
Catalog No. AA4688404
Description
Description
  • Calayst for hydrogen transfers including reduction of ketones and aldehydes, and oxidation and/or racemization of amines and alcohols
Specifications
Specifications
1-Hydroxytetraphenylcyclopentadienyl (tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldiruthenium (II)
Yellow to Orange
2g
C62H42O6Ru2
14,8483
1085.15
98%
Moisture Sensitive
223°C to 227°C
104439-77-2
MFCD03840556
Shvo's Catalyst
Crystalline
Chemical Identifiers
104439-77-2
C62H42O6Ru2
MFCD03840556
Shvo's Catalyst
SDS Documents