missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(Hydroxymethyl)-5,5-dimethylhydantoin 98.0+%, TCI America™
$54.75 - $336.78
Chemical Identifiers
CAS | 116-25-6 |
---|---|
Molecular Formula | C6H10N2O3 |
Molecular Weight (g/mol) | 158.16 |
MDL Number | MFCD00020866 |
InChI Key | SIQZJFKTROUNPI-UHFFFAOYSA-N |
Synonym | 1-hydroxymethyl-5,5-dimethylhydantoin, mdm hydantoin, glycoserve, mdmh, 1-hydroxymethyl-5,5-dimethylimidazolidine-2,4-dione, mdm hydantoin van, monomethyloldimethylhydantoin, 1-monomethylol-5,5-dimethylhydantoin, methylol dimethylhydantoin |
PubChem CID | 67000 |
IUPAC Name | 1-(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione |
SMILES | CC1(C)N(CO)C(=O)NC1=O |
Chemical Identifiers
116-25-6 | |
158.16 | |
SIQZJFKTROUNPI-UHFFFAOYSA-N | |
67000 | |
CC1(C)N(CO)C(=O)NC1=O |
C6H10N2O3 | |
MFCD00020866 | |
1-hydroxymethyl-5,5-dimethylhydantoin, mdm hydantoin, glycoserve, mdmh, 1-hydroxymethyl-5,5-dimethylimidazolidine-2,4-dione, mdm hydantoin van, monomethyloldimethylhydantoin, 1-monomethylol-5,5-dimethylhydantoin, methylol dimethylhydantoin | |
1-(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione |
Specifications
116-25-6 | |
C6H10N2O3 | |
25 g | |
SIQZJFKTROUNPI-UHFFFAOYSA-N | |
1-(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione | |
67000 | |
≥98.0% (HPLC,T) | |
1-(Hydroxymethyl)-5,5-dimethylhydantoin |
White | |
MFCD00020866 | |
1-hydroxymethyl-5,5-dimethylhydantoin, mdm hydantoin, glycoserve, mdmh, 1-hydroxymethyl-5,5-dimethylimidazolidine-2,4-dione, mdm hydantoin van, monomethyloldimethylhydantoin, 1-monomethylol-5,5-dimethylhydantoin, methylol dimethylhydantoin | |
CC1(C)N(CO)C(=O)NC1=O | |
158.16 | |
158.16 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3543
TSCA : Yes