Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Hydroxymethyl-5,5-dimethylhydantoin, 97%

$93.73 - $301.35

Chemical Identifiers

Chemical Name or Material 1-Hydroxymethyl-5,5-dimethylhydantoin
CAS 116-25-6
Molecular Formula C6H10N2O3
Formula Weight 158.16
MDL Number MFCD00020866
Beilstein 139160
Merck Index 14,4831
Synonym 5,5-Dimethyl-1-(hydroxymethyl)hydantoin
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1986814

  SDS
alfa aesar™
A1986814
25g Each for $93.73

AAA1986822

  SDS
alfa aesar™
A1986822
100g Each for $301.35
Description & Specifications

Specifications

Formula Weight 158.16
Chemical Name or Material 1-Hydroxymethyl-5,5-dimethylhydantoin
TSCA TSCA
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 25g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C6H10N2O3
CAS 116-25-6
Synonym 5,5-Dimethyl-1-(hydroxymethyl)hydantoin
MDL Number MFCD00020866
Melting Point 104° to 106°C
Merck Index 14,4831
Beilstein 139160
Health Hazard 1 Warning
RTECS Number NI9454200

Applications
1-Hydroxymethyl-5,5-dimethylhydantoin is used as chelator for light-sensitive pharmaceutical drug, and as raw material of water-soluble polymers. It also can be used as raw material of disinfectants and preservatives, and soluble inhibitor for proteins. It is used as an adjuvant in the bleaching of food-contact recycled paper and board used in food packaging.

Solubility
Freely soluble in water.

Notes
Moisture Sensitive. Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.