Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Hydroxyisoquinoline, 97%

$121.91 - $629.82

Chemical Identifiers

Chemical Name or Material 1-Hydroxyisoquinoline
CAS 491-30-5
Molecular Formula C9H7NO
Formula Weight 145.16
MDL Number MFCD00006899
Synonym 1-Isoquinolinol; Isocarbostyril
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2912403

  SDS
alfa aesar™
H2912403
1g Each for $121.91

AAH2912409

  SDS
alfa aesar™
H2912409
10g Each for $629.82
Description & Specifications

Specifications

Formula Weight 145.16
CAS 491-30-5
Quantity 1g
Melting Point 212° to 215°C
Health Hazard 1 Warning
Molecular Formula C9H7NO
MDL Number MFCD00006899
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 97%
Chemical Name or Material 1-Hydroxyisoquinoline
Synonym 1-Isoquinolinol; Isocarbostyril
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.