Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Hydroxyethylidenebis(phosphonic acid), 96%

$22.57 - $22.57

Chemical Identifiers

Chemical Name or Material 1-Hydroxyethylidenebis (phosphonic acid)
CAS 2809-21-4
Molecular Formula CH3C(OH)(PO3H2)2
Formula Weight 206.03
MDL Number MFCD00070585
UN Number UN3261
Merck Index 14,3863
Synonym Etidronic acid; 1-Hydroxyethane-1,1-diphosphonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4350204

  SDS
alfa aesar™
4350204
2g Each for $22.57

AA4350209

  SDS
alfa aesar™
4350209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 206.03
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1-Hydroxyethylidenebis (phosphonic acid)
TSCA TSCA
Physical Form Powder
Assay 96%
Molecular Formula CH3C(OH)(PO3H2)2
MDL Number MFCD00070585
CAS 2809-21-4
UN Number UN3261
Merck Index 14,3863
Synonym Etidronic acid; 1-Hydroxyethane-1,1-diphosphonic acid
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECS Number SZ8562100
Odor Odorless
Quantity 2g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.