missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Hydroxyethylidenebis(phosphonic acid), 96%

Manufacturer:  Alfa Aesar™ 4350218

 View more versions of this product

Catalog No. AA4350218



Specifications

Specifications

1-Hydroxyethylidenebis (phosphonic acid)
50g
C2H8O7P2
UN3261
1-hydroxyethylidene-1,1-diphosphonic acid
CC(O)(P(=O)(O)O)P(=O)(O)O
206.027
CHEBI:4907
Powder
Odorless
2809-21-4
MFCD00070585
14,3863
DBVJJBKOTRCVKF-UHFFFAOYSA-N
(1-hydroxy-1-phosphonoethyl)phosphonic acid
3305
206.03
96%

Chemical Identifiers

2809-21-4
206.027
DBVJJBKOTRCVKF-UHFFFAOYSA-N
3305
(1-hydroxy-1-phosphonoethyl)phosphonic acid
C2H8O7P2
MFCD00070585
1-hydroxyethylidene-1,1-diphosphonic acid
CHEBI:4907
CC(O)(P(=O)(O)O)P(=O)(O)O
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : SZ8562100

TSCA : TSCA

Documents