missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Hydroxyethylethoxypiperazine, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 259110050
Specifications
1-Hydroxyethylethoxypiperazine | |
Authentic | |
1.4960 to 1.4980 | |
150.0°C (13.0 mmHg) | |
5mL | |
94.0 | |
94% min. (GC) | |
MFCD00074931 | |
FLNQAPQQAZVRDA-UHFFFAOYSA-N | |
2-(2-piperazin-1-ylethoxy)ethanol | |
139436 | |
Liquid |
1.0100g/mL | |
Glass bottle | |
1.01 | |
Brown-Yellow to Colorless | |
13349-82-1 | |
100.0 | |
C8H18N2O2 | |
1-2-2-hydroxyethoxy ethyl piperazine, 1-2-2-hydroxyethoxy-ethyl-piperazine, 1-2-2-hydroxyethoxyl-ethyl piperazine, 2-2-1-piperazinyl ethoxy ethanol, 2-2-piperazin-1-yl ethoxy ethanol, 2-2-piperazin-1-ylethoxy ethanol, ethanol, 2-2-1-piperazinyl ethoxy | |
C1CN(CCN1)CCOCCO | |
174.24 | |
174.24 | |
95% |
Chemical Identifiers
13349-82-1 | |
174.24 | |
FLNQAPQQAZVRDA-UHFFFAOYSA-N | |
139436 | |
C1CN(CCN1)CCOCCO |
C8H18N2O2 | |
MFCD00074931 | |
1-2-2-hydroxyethoxy ethyl piperazine, 1-2-2-hydroxyethoxy-ethyl-piperazine, 1-2-2-hydroxyethoxyl-ethyl piperazine, 2-2-1-piperazinyl ethoxy ethanol, 2-2-piperazin-1-yl ethoxy ethanol, 2-2-piperazin-1-ylethoxy ethanol, ethanol, 2-2-1-piperazinyl ethoxy | |
2-(2-piperazin-1-ylethoxy)ethanol |