missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Hydroxy-2-naphthoic Acid, TCI America™

$25.97 - $148.20

Chemical Identifiers

CAS 86-48-6
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD00003960
InChI Key SJJCQDRGABAVBB-UHFFFAOYSA-N
Synonym 1-hydroxy-2-naphthoic acid, 1-naphthol-2-carboxylic acid, 2-carboxy-1-naphthol, 2-naphthalenecarboxylic acid, 1-hydroxy, 1-hydroxy-2-naphthoate, 1-hydroxy-2-naphthalenecarboxylic acid, unii-u8lz3r07l8, 2-naphthoic acid, 1-hydroxy, alpha-hydroxynaphthoic acid, 1-hydroxy-naphthalene-2-carboxylic acid
PubChem CID 6844
ChEBI CHEBI:36108
IUPAC Name 1-hydroxynaphthalene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C=CC(=C2O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H079625G View Documents TCI America
H079625G
25g Each for $25.97
 
H0796500G View Documents TCI America
H0796500G
500g Each for $148.20
 
Specifications

Chemical Identifiers

86-48-6
188.182
SJJCQDRGABAVBB-UHFFFAOYSA-N
6844
1-hydroxynaphthalene-2-carboxylic acid
C11H8O3
MFCD00003960
1-hydroxy-2-naphthoic acid, 1-naphthol-2-carboxylic acid, 2-carboxy-1-naphthol, 2-naphthalenecarboxylic acid, 1-hydroxy, 1-hydroxy-2-naphthoate, 1-hydroxy-2-naphthalenecarboxylic acid, unii-u8lz3r07l8, 2-naphthoic acid, 1-hydroxy, alpha-hydroxynaphthoic acid, 1-hydroxy-naphthalene-2-carboxylic acid
CHEBI:36108
C1=CC=C2C(=C1)C=CC(=C2O)C(=O)O

Specifications

Undesignated
25g
C11H8O3
1-hydroxy-2-naphthoic acid, 1-naphthol-2-carboxylic acid, 2-carboxy-1-naphthol, 2-naphthalenecarboxylic acid, 1-hydroxy, 1-hydroxy-2-naphthoate, 1-hydroxy-2-naphthalenecarboxylic acid, unii-u8lz3r07l8, 2-naphthoic acid, 1-hydroxy, alpha-hydroxynaphthoic acid, 1-hydroxy-naphthalene-2-carboxylic acid
C1=CC=C2C(=C1)C=CC(=C2O)C(=O)O
188.182
CHEBI:36108
Crystal-Powder at 20°C
1-Hydroxy-2-naphthoic Acid
195°C
86-48-6
MFCD00003960
SJJCQDRGABAVBB-UHFFFAOYSA-N
1-hydroxynaphthalene-2-carboxylic acid
6844
188.18
≥98.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0398

TSCA : Yes