Please login to your online account to display your discounted pricing

1-Hydroxy-2-naphthoic acid, 98%, ACROS Organics™

$19.52 - $19.52

Chemical Identifiers

Chemical Name or Material 1-Hydroxy-2-naphthoic acid
Name Note 99%
CAS 86-48-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥97.5% (Acid)
Assay Percent Range 98%
Molecular Formula C11H8O3
Formula Weight 188.18
MDL Number MFCD00003960
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC149230050

  SDS
thermo fisher scientific
149230050
Glass bottle 5g Each for $19.52

AC149231000

  SDS
fisher scientific acros
149231000
Plastic bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Crystals
Assay ≥97.5% (Acid)
Formula Weight 188.18
Color White to slightly pink to tan or
Melting Point 195° to 205°C (dec.)
Quantity 5g
EINECS Number 201-674-0
TSCA TSCA
Beilstein 10, 331
Merck Index 15, 4873
CAS Max % 100.0
Chemical Name or Material 1-Hydroxy-2-naphthoic acid
Packaging Glass bottle
Name Note 99%
Assay Percent Range 98%
Molecular Formula C11H8O3
MDL Number MFCD00003960
CAS 86-48-6
CAS Min % 98.5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.