Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Hydroxy-2-naphthoic acid, 98+%

$72.18 - $235.05

Chemical Identifiers

Chemical Name or Material 1-Hydroxy-2-naphthoic acid
CAS 86-48-6
Molecular Formula C11H8O3
Formula Weight 188.18
MDL Number MFCD00003960
Beilstein 974438
Merck Index 14,4834
Synonym 1-Naphthol-2-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA127850B

  SDS
alfa aesar™
A127850B
1000g Each for $235.05

AAA1278530

  SDS
alfa aesar™
A1278530
250g Each for $72.18

AAA1278536

  SDS
alfa aesar™
A1278536
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 1-Hydroxy-2-naphthoic acid
Melting Point ≈200°C (dec.)
Quantity 1000g
CAS 86-48-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay ≥98%
Molecular Formula C11H8O3
Formula Weight 188.18
MDL Number MFCD00003960
Beilstein 974438
Merck Index 14,4834
Synonym 1-Naphthol-2-carboxylic acid

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Soluble in alcohol, ethanol, benzene, methanol, and water (partly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.