Learn More
1-Hexyne, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1315609
Description
1-Hexyne is used in the preparation of tricyclic isoindolinone scaffold by undergoing hydrozirconation and ring-closing metathesis. It is used to prepare trans-1,2-bis(5-thianthreniumyl)alkene tetrafluoroborate by reacting with thianthrene cation radical tetrafluoroborate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Hexyne is used in the preparation of tricyclic isoindolinone scaffold by undergoing hydrozirconation and ring-closing metathesis. It is used to prepare trans-1,2-bis(5-thianthreniumyl)alkene tetrafluoroborate by reacting with thianthrene cation radical tetrafluoroborate.
Solubility
Miscible with organic solvents. Immiscible with water.
Notes
Incompatible with strong oxidizing agents, strong reducing agents, strong acids and strong bases.
Specifications
1-Hexyne | |
-132°C | |
69°C to 71°C | |
Characteristic | |
1.399 | |
10 g | |
635687 | |
Miscible with organic solvents. Immiscible with water. | |
CCCCC#C | |
82.146 | |
82.15 |
693-02-7 | |
0.718 | |
−21°C (−5°F) | |
C6H10 | |
MFCD00009504 | |
UN3295 | |
1-hexyne, n-butylacetylene, butylacetylene, hexyne, unii-5fzf2f38f5, n-hexyne, 1-hexyn, hex-5-yn-1-yl, pubchem13057 | |
CGHIBGNXEGJPQZ-UHFFFAOYSA-N | |
hex-1-yne | |
12732 | |
≥98% |
Chemical Identifiers
693-02-7 | |
82.146 | |
CGHIBGNXEGJPQZ-UHFFFAOYSA-N | |
12732 | |
CCCCC#C |
C6H10 | |
MFCD00009504 | |
1-hexyne, n-butylacetylene, butylacetylene, hexyne, unii-5fzf2f38f5, n-hexyne, 1-hexyn, hex-5-yn-1-yl, pubchem13057 | |
hex-1-yne |
Safety and Handling
GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 211-736-9
RTECSNumber : MQ9691000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only