Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Hexyn-3-ol, 97%

Chemical Identifiers

Chemical Name or Material 1-Hexyn-3-ol
CAS 105-31-7
Molecular Formula C6H10O
Formula Weight 98.15
MDL Number MFCD00014408
UN Number UN2929
Beilstein 1739419
Synonym Ethynyl n-propyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1154906

 
alfa aesar™
L1154906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1154914

 
alfa aesar™
L1154914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 98.15
Chemical Name or Material 1-Hexyn-3-ol
TSCA TSCA
Assay 97%
Molecular Formula C6H10O
CAS 105-31-7
UN Number UN2929
Beilstein 1739419
Synonym Ethynyl n-propyl carbinol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H310-H226
Toxic if swallowed.
Fatal in contact with skin.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number MR0181000
Boiling Point 142° to 143°C
Quantity 5g
Odor Recognizable
MDL Number MFCD00014408
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
Flash Point 47°C (116°F)
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H310-H226
Toxic if swallowed.
Fatal in contact with skin.
Flammable liquid and vapor.