missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Hexyn-3-ol, 97%

Manufacturer:  Alfa Aesar™ L1154914

 View more versions of this product

Catalog No. AAL1154914



Specifications

Specifications

1-Hexyn-3-ol
Recognizable
25g
C6H10O
UN2929
1-hexyn-3-ol, propylethinylcarbinol, 1-hexyne-3-ol, 3-hydroxy-1-hexyne, 1-propylpropargyl alcohol, 1-hexine-3-ol, acmc-2098fm, dsstox_cid_30088, dsstox_gsid_51536, 4-01-00-02234 beilstein handbook reference
CCCC(C#C)O
98.145
98.15
47°C (116°F)
142°C to 143°C
105-31-7
MFCD00014408
1739419
LTFTWJYRQNTCHI-UHFFFAOYSA-N
hex-1-yn-3-ol
7746
97%

Chemical Identifiers

105-31-7
98.145
LTFTWJYRQNTCHI-UHFFFAOYSA-N
7746
CCCC(C#C)O
C6H10O
MFCD00014408
1-hexyn-3-ol, propylethinylcarbinol, 1-hexyne-3-ol, 3-hydroxy-1-hexyne, 1-propylpropargyl alcohol, 1-hexine-3-ol, acmc-2098fm, dsstox_cid_30088, dsstox_gsid_51536, 4-01-00-02234 beilstein handbook reference
hex-1-yn-3-ol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H310-H226
Toxic if swallowed.
Fatal in contact with skin.
Flammable liquid and vapor.

GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.

RTECSNumber : MR0181000

TSCA : TSCA

Documents