missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Hexylpyridinium Hexafluorophosphate, TCI America™

$282.64 - $965.58

Chemical Identifiers

CAS 797789-00-5
Molecular Formula C11H18F6NP
Molecular Weight (g/mol) 309.236
MDL Number MFCD03093298
InChI Key DKRTWJPQQYAIHH-UHFFFAOYSA-N
Synonym 1-hexylpyridinium hexafluorophosphate, 1-hexylpyridin-1-ium hexafluorophosphate v, 1-hexylpyridin-1-ium hexafluorophosphate, dsstox_cid_27912, dsstox_rid_82665, dsstox_gsid_47936, ksc376g7d, n-hexylpyridinium hexafluorophosphate
PubChem CID 2734177
IUPAC Name 1-hexylpyridin-1-ium;hexafluorophosphate
SMILES CCCCCC[N+]1=CC=CC=C1.F[P-](F)(F)(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H14245G View Documents TCI America
H14245G
5g Each for $282.64
 
H142425G View Documents TCI America
H142425G
25g Each for $965.58
 
Specifications

Chemical Identifiers

797789-00-5
309.236
DKRTWJPQQYAIHH-UHFFFAOYSA-N
2734177
CCCCCC[N+]1=CC=CC=C1.F[P-](F)(F)(F)(F)F
C11H18F6NP
MFCD03093298
1-hexylpyridinium hexafluorophosphate, 1-hexylpyridin-1-ium hexafluorophosphate v, 1-hexylpyridin-1-ium hexafluorophosphate, dsstox_cid_27912, dsstox_rid_82665, dsstox_gsid_47936, ksc376g7d, n-hexylpyridinium hexafluorophosphate
1-hexylpyridin-1-ium;hexafluorophosphate

Specifications

White
5g
C11H18F6NP
1-hexylpyridinium hexafluorophosphate, 1-hexylpyridin-1-ium hexafluorophosphate v, 1-hexylpyridin-1-ium hexafluorophosphate, dsstox_cid_27912, dsstox_rid_82665, dsstox_gsid_47936, ksc376g7d, n-hexylpyridinium hexafluorophosphate
CCCCCC[N+]1=CC=CC=C1.F[P-](F)(F)(F)(F)F
309.236
309.24
≥98.0% (HPLC,T)
48°C
797789-00-5
MFCD03093298
DKRTWJPQQYAIHH-UHFFFAOYSA-N
1-hexylpyridin-1-ium;hexafluorophosphate
2734177
Crystal-Powder at 20°C
1-Hexylpyridinium Hexafluorophosphate
Safety and Handling

Safety and Handling

TSCA : No