missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Hexyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ A1222306

 View more versions of this product

Catalog No. AAA1222306



Specifications

Specifications

1-Hexyl isothiocyanate
Pungent
5g
C7H13NS
UN2922
hexyl isothiocyanate, n-hexyl isothiocyanate, hexane, 1-isothiocyanato, 1-hexyl isothiocyanate, hexylisothiocyanate, isothiocyanic acid, hexyl ester, unii-2s7c3ga0z5, 1-hexylisothiocyanate, 1-isothiocyanato-hexane, hexanisothiocyanate
CCCCCCN=C=S
143.248
143.25
90°C (194°F)
216°C
4404-45-9
MFCD00014445
1751837
WXYAXKKXIGHXDS-UHFFFAOYSA-N
1-isothiocyanatohexane
78120
97%

Chemical Identifiers

4404-45-9
143.248
WXYAXKKXIGHXDS-UHFFFAOYSA-N
78120
CCCCCCN=C=S
C7H13NS
MFCD00014445
hexyl isothiocyanate, n-hexyl isothiocyanate, hexane, 1-isothiocyanato, 1-hexyl isothiocyanate, hexylisothiocyanate, isothiocyanic acid, hexyl ester, unii-2s7c3ga0z5, 1-hexylisothiocyanate, 1-isothiocyanato-hexane, hexanisothiocyanate
1-isothiocyanatohexane
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

SDS Documents