Learn More
1-Hexyl-3-methylimidazolium hexafluorophosphate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 354181000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Ionic liquidSpecifications
1-Hexyl-3-methylimidazolium hexafluorophosphate | |
98.5 | |
1.3045g/mL | |
99% | |
C10H19N2·PF6 | |
MFCD03093296 | |
1.3045 | |
YPWSSSRXUOQNMQ-UHFFFAOYSA-N | |
1-hexyl-3-methylimidazol-3-ium;hexafluorophosphate | |
2734175 | |
312.24 |
304680-35-1 | |
100.0 | |
Authentic | |
Plastic bottle | |
1.4160 to 1.418 | |
100g | |
1-hexyl-3-methylimidazolium hexafluorophosphate, 1-hexyl-3-methyl-1h-imidazol-3-ium hexafluorophosphate v, 1-n-hexyl-3-methylimidazolium hexafluorophosphate, acmc-209hg2, dsstox_cid_27925, dsstox_rid_82678, dsstox_gsid_47949, ksc222c9l, 1-hexyl-3-methyl-imidazol-3-ium hexafluorophosphate, 1-hex-1-yl-3-methyl-1h-imidazol-3-ium hexafluorophosphate | |
CCCCCCN1C=C[N+](=C1)C.F[P-](F)(F)(F)(F)F | |
312.24 | |
497 mPa.s (25°C) | |
99% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
TSCA : Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.
RUO â Research Use Only