missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Hexene, 99%, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC120751000

 View more versions of this product

Catalog No. AC120751000



Description

Description

Specifications

Specifications

1-Hexene
−26°C
AcroSeal™ Glass Bottle
0.678
62.0°C to 65.0°C
-140.0°C
592-41-6
100.0
C6H12
MFCD00009505
1-hexene, 1-n-hexene, butyl ethylene, butylethylene, dialene 6, hexene, hexene-1, hexylene, n-hexene, unii-b38zz8c206
LIKMAJRDDDTEIG-UHFFFAOYSA-N
hex-1-ene
11597
84.15
99%
0.6780g/mL
Authentic
1.3867 to 1.3887
0.34 mm2/s (40°C)
Colorless
100mL
98.5
98.5% min. (GC)
CH3(CH2)3CH=CH2
01,215
Solubility in water: 50 mg/l (20°C). Other solubilities: soluble in alcohol and hydrocarbon solvents, soluble in ethanol and diethyl ether
CCCCC=C
84.15
CHEBI:24579
Liquid

Chemical Identifiers

592-41-6
84.15
LIKMAJRDDDTEIG-UHFFFAOYSA-N
11597
hex-1-ene
C6H12
MFCD00009505
1-hexene, 1-n-hexene, butyl ethylene, butylethylene, dialene 6, hexene, hexene-1, hexylene, n-hexene, unii-b38zz8c206
CHEBI:24579
CCCCC=C
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take precautionary measures against static discharge.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Imm

GHS Signal Word: Danger

EINECSNumber : 209-753-1

SDS Documents