missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Hexene, 97%, Thermo Scientific Chemicals

1-Hexene, 97%, C6H12, CAS Number-592-41-6, dialene 6, butyl ethylene, 1-hexene, butylethylene, 1-n-hexene, hexylene, hexene-1, hexene, n-hexene, unii-b38zz8c206, 1L, 96% min. (GC), 01, 215, 62.0 deg.C to 65.0 deg.C, 100.0, 96.0, CHEBI:24579, Colorless, 0.6780g/mL, 209-753-1, -26 deg.C, 84.15

Supplier:  Thermo Scientific Chemicals 213210010

 View more versions of this product

Catalog No. AC213210010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Hexene
96.0
-140.0°C
0.6780g/mL
−26°C
96% min. (GC)
C6H12
CH3(CH2)3CH=CH2
1 L
0.678
Solubility in water: 50 mg/l (20°C). Other solubilities: soluble in alcohol and hydrocarbon solvents, soluble in ethanol and diethyl ether
CCCCC=C
84.15
0.34 mm2/s (40°C)
84.15
Liquid
592-41-6
100.0
Colorless
62.0°C to 65.0°C
Authentic
Glass bottle
1.3867 to 1.3887
MFCD00009505
01, 215
1-hexene, hexene, butylethylene, hexylene, hexene-1, 1-n-hexene, butyl ethylene, dialene 6, n-hexene, unii-b38zz8c206
LIKMAJRDDDTEIG-UHFFFAOYSA-N
hex-1-ene
11597
CHEBI:24579
97%

Chemical Identifiers

592-41-6
84.15
LIKMAJRDDDTEIG-UHFFFAOYSA-N
11597
hex-1-ene
C6H12
MFCD00009505
1-hexene, hexene, butylethylene, hexylene, hexene-1, 1-n-hexene, butyl ethylene, dialene 6, n-hexene, unii-b38zz8c206
CHEBI:24579
CCCCC=C
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take precautionary measures against static discharge.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Imm

GHS Signal Word: Danger

EINECSNumber : 209-753-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only