Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Hexen-3-ol, 98%

Chemical Identifiers

Chemical Name or Material 1-Hexen-3-ol
CAS 4798-44-1
Molecular Formula C6H12O
Formula Weight 100.16
MDL Number MFCD00004581
UN Number UN1987
Beilstein 1720166
Synonym n-Propyl vinyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1967209

  SDS
alfa aesar™
A1967209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1967218

  SDS
alfa aesar™
A1967218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 100.16
Chemical Name or Material 1-Hexen-3-ol
TSCA TSCA
Assay 98%
Molecular Formula C6H12O
MDL Number MFCD00004581
CAS 4798-44-1
UN Number UN1987
Beilstein 1720166
Synonym n-Propyl vinyl carbinol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H302
Flammable liquid and vapour.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Flash Point 35°C (95°F)
Odor Characteristic
Boiling Point 132° to 134°C
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H302
Flammable liquid and vapour.
Harmful if swallowed.