missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Hexanethiol, 97%

Manufacturer:  Alfa Aesar™ L09659AE

 View more versions of this product

Catalog No. AAL09659AE



Description

Description

Applications
It is used to tune the electrical characteristics in networked carbon nanotube field-effect transistors.1 Also used as a monolayer on copper surfaces to retard surface oxidation during exposure to the environment which can then be desorbed effectively with an annealing step in inert N2 to provide a clean Cu surface.

Solubility
Difficult to mix in water.

Notes
Air sensitive. Store in cool dry condition. Incompatible with strong oxidizing agents.
Specifications

Specifications

1-Hexanethiol
Strong
-81°C
111-31-9
CH3(CH2)5SH
UN3071
1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan
CCCCCCS
118.238
118.24
30°C (86°F)
150°C to 154°C
100mL
C6H14S
MFCD00004909
1731295
PMBXCGGQNSVESQ-UHFFFAOYSA-N
hexane-1-thiol
8106
97%

Chemical Identifiers

111-31-9
118.238
PMBXCGGQNSVESQ-UHFFFAOYSA-N
8106
CCCCCCS
C6H14S
MFCD00004909
1-hexanethiol, hexyl mercaptan, n-hexyl mercaptan, hexanethiol, 1-hexylthiol, 1-mercaptohexane, hexylthiol, n-hexanethiol, n-hexylthiol, n-hexylmercaptan
hexane-1-thiol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H226
Toxic if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.

GHS P Statement
P260-P280f-P309-P310-P403
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a well-ventilated place.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCAPTANS, LIQUID, TOXIC, FLAMMABLE, N.O.S.

RTECSNumber : MO4550000

TSCA : TSCA

Documents