missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Hexanesulfonic Acid, Sodium Salt, Monohydrate, HPLC Grade, 99% min., EMD Millipore

Manufacturer:  Millipore Sigma HX01022

 View more versions of this product

Catalog No. MHX01022



Specifications

Specifications

1-Hexanesulfonic Acid, Sodium Salt, Monohydrate
LiChropur™, For Ion Pair Chromatography
99% min.
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
CCCCCCS(=O)(=O)[O-].[Na+]
188.217
188.22
25g
2832-45-3
C6H13NaO3S
QWSZRRAAFHGKCH-UHFFFAOYSA-M
sodium;hexane-1-sulfonate
23677630

Chemical Identifiers

2832-45-3
188.217
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
sodium;hexane-1-sulfonate
C6H13NaO3S
QWSZRRAAFHGKCH-UHFFFAOYSA-M
23677630
CCCCCCS(=O)(=O)[O-].[Na+]
Safety and Handling

Safety and Handling

Recommended Storage : Store at +5°C to +30°C

Documents