Learn More
1-Hexanesulfonic acid, sodium salt hydrate, 98%, HPLC grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 411600050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatographySpecifications
1-Hexanesulfonic acid, sodium salt hydrate | |
(0.25M aq. soln.) (1cm cell vs HPLC grade water), 0.2 max. at 210nm, 0.1 max. at 220nm, 0.075 max. at 230nm, 0.05 max. at 240nm, 0.05 max. at 250nm, 0.04 max. at 254nm, 0.04 max. at 280nm, (0.25 M aq. soln.) (1 cm cell vs HPLC grade water), (0.25 M aq. soln.) (1 cm cell vs HPLC grade water) at 0nm | |
Colorless to Yellow | |
Authentic | |
Glass bottle | |
CH3(CH2)5SO3Na·xH2O | |
5 g | |
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt | |
QWSZRRAAFHGKCH-UHFFFAOYSA-M | |
sodium;hexane-1-sulfonate | |
23677630 | |
Negligible | |
Liquid |
2832-45-3 | |
>300°C | |
Negligible | |
98% | |
C6H13NaO3S | |
MFCD00007542 | |
04, IV, 54 | |
(10% in water) Clear colorless | |
[Na+].CCCCCCS([O-])(=O)=O | |
188.22 | |
188.22 | |
98% |
Chemical Identifiers
2832-45-3 | |
188.22 | |
QWSZRRAAFHGKCH-UHFFFAOYSA-M | |
23677630 | |
[Na+].CCCCCCS([O-])(=O)=O |
C6H13NaO3S | |
MFCD00007542 | |
sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt | |
sodium;hexane-1-sulfonate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
GHS Signal Word: Warning
RUO – Research Use Only