Please login to your online account to display your discounted pricing

1-Hexanesulfonic Acid Sodium Salt, HPLC Grade, J.T.Baker™

High purity solvents for use in analytical and preparative separations

$285.97 - $3,952.85

Chemical Identifiers

Chemical Name or Material 1-Hexanesulfonic Acid Sodium Salt
CAS 2832-45-3
Purity Grade HPLC Grade
Molecular Formula CH3(CH2)5SO3Na
Formula Weight 188.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002042

 
avantor performance
217505
Wide Mouth Amber Glass Bottle 25g Each for $285.97

02002043

 
avantor performance
217501
Wide Mouth Amber Glass Bottle 500g Each for $3,952.85
Description & Specifications

Specifications

Formula Weight 188.22
Chemical Name or Material 1-Hexanesulfonic Acid Sodium Salt
Molecular Formula CH3(CH2)5SO3Na
CAS 2832-45-3
Purity Grade HPLC Grade
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 0.00kg
Quantity 25g

    For Ion-Pair Chromatography of Basic Compounds