Please login to your online account to display your discounted pricing

1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-Hexadecylpyridinium chloride monohydrate
Name Note 96.0-101.0%
CAS 6004-24-6
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0 to 101.0%
Assay Percent Range 96.0-101.0%
Molecular Formula C21H38ClN·H2O
Formula Weight 358.01
MDL Number MFCD00149977
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC226990025

  SDS
thermo fisher scientific
AC226990025
2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 358.01
Quantity 2.5kg
Color White
Assay 96.0 to 101.0%
RTECS Number UU5075000
Merck Index 15, 2031
CAS Min % 96.0
CAS Max % 100.0
Beilstein 20, V,5, 233
Chemical Name or Material 1-Hexadecylpyridinium chloride monohydrate
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Water 4.5 to 5.5%
Name Note 96.0-101.0%
Assay Percent Range 96.0-101.0%
Molecular Formula C21H38ClN·H2O
MDL Number MFCD00149977
CAS 6004-24-6
Synonym Cetylpyridinium chloride monohydrate
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Fatal if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Fatal if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.