Please login to your online account to display your discounted pricing

Alfa Aesar™ (1-Hexadecyl)pyridinium bromide monohydrate, 98%

$38.18 - $491.03

Chemical Identifiers

Chemical Name or Material (1-Hexadecyl)pyridinium bromide monohydrate
CAS 140-72-7
Molecular Formula C21H38BrN·H2O
Formula Weight 402.47 (384.44 Anhydrous)
MDL Number MFCD00150003
Beilstein 3778948
Synonym Cetylpyridinium bromide monohydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA148250B

 
alfa aesar™
A148250B
1000g Each for $491.03

AAA1482518

 
alfa aesar™
A1482518
50g Each for $38.18

AAA1482530

 
alfa aesar™
A1482530
250g Each for $142.24
Description & Specifications

Specifications

Formula Weight 402.47 (384.44 Anhydrous)
Chemical Name or Material (1-Hexadecyl)pyridinium bromide monohydrate
TSCA TSCA
Assay 98%
Molecular Formula C21H38BrN·H2O
MDL Number MFCD00150003
CAS 140-72-7
Beilstein 3778948
Synonym Cetylpyridinium bromide monohydrate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UU4848000
Melting Point 67° to 69°C
Quantity 1000g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.