missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Hexadecene, 90+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L02637AK

 View more versions of this product

Catalog No. AAL02637AK



Description

Description

1-Hexadecene is used as an intermediate for organic synthesis. It is utilized as a blend for off-shore drilling fluids. It is also used as a surface active agent, finishing agent, process regulator, functional fluid, lubricant and lubricant additive. Further, it acts as comonomers in polymer such as low density polyethylenes, which shows thiner, flexibility and tearing resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexadecene is used as an intermediate for organic synthesis. It is utilized as a blend for off-shore drilling fluids. It is also used as a surface active agent, finishing agent, process regulator, functional fluid, lubricant and lubricant additive. Further, it acts as comonomers in polymer such as low density polyethylenes, which shows thiner, flexibility and tearing resistance.

Solubility
Miscible with ether, petroleum ether and alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

1-Hexadecene
4°C
273°C to 274°C
C16H32
MFCD00008991
1753401
Miscible with ether,petroleum ether and alcohol. Immiscible with water.
CCCCCCCCCCCCCCC=C
224.432
CHEBI:77507
94%
629-73-2
0.781
132°C (269°F)
1.442
250 mL
1-hexadecene, cetene, 1-cetene, hexadecylene-1, hexadecene, 1-n-hexadecene, cetylene, n-hexadec-1-ene, alpha-hexadecene, alpha-hexadecylene
GQEZCXVZFLOKMC-UHFFFAOYSA-N
hexadec-1-ene
12395
224.43

Chemical Identifiers

629-73-2
224.432
GQEZCXVZFLOKMC-UHFFFAOYSA-N
12395
hexadec-1-ene
C16H32
MFCD00008991
1-hexadecene, cetene, 1-cetene, hexadecylene-1, hexadecene, 1-n-hexadecene, cetylene, n-hexadec-1-ene, alpha-hexadecene, alpha-hexadecylene
CHEBI:77507
CCCCCCCCCCCCCCC=C
Safety and Handling

Safety and Handling

P301+P310-P331-P501c

H304

EINECSNumber : 211-105-8

RTECSNumber : MM0479000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only