missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Heptyne, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 223461000

 View more versions of this product

Catalog No. AC223461000


 

Specifications Description & Specifications

Specifications

1-Heptyne
99%
98.5
Authentic
≥98.5%
C7H12
CH3(CH2)4C2CH
MFCD00009529
1.4070 to 1.4090 (20°C, 589nm)
Clear colorless to light yellow-green
100mL
628-71-7
100.0
Liquid
99%
Glass bottle
96.17
01, 256
0.73g/mL

Chemical Identifiers

628-71-7
100.0
C7H12
96.17
1-Heptyne, 99%
98.5
99%
CH3(CH2)4C2CH
MFCD00009529
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-051-5

TSCA : TSCA