missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Heptyne, 99%

Manufacturer:  Alfa Aesar™ A1113022

 View more versions of this product

Catalog No. AAA1113022



Description

Description

Applications
1-Heptyne is used in the synthesis of (6Z,10E,12Z)-octadeca-6,10,12-trienoic acid and (8Z,12E,14Z)-eicosa-8,12,14-trienoic acid.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

1-Heptyne
Unpleasant
-81°C
628-71-7
MFCD00009529
1733518
YVXHZKKCZYLQOP-UHFFFAOYSA-N
hept-1-yne
12350
99%
−10°C (14°F)
98°C to 100°C
100g
C7H12
UN3295
1-heptyne, amylacetylene, heptyne, unii-z3nf9ssh6k, z3nf9ssh6k, amyl acetylene, n-pentylacetylene, pubchem13058, 1-heptyne, acmc-1aw2o
CCCCCC#C
96.173
96.17

Chemical Identifiers

628-71-7
96.173
YVXHZKKCZYLQOP-UHFFFAOYSA-N
12350
CCCCCC#C
C7H12
MFCD00009529
1-heptyne, amylacetylene, heptyne, unii-z3nf9ssh6k, z3nf9ssh6k, amyl acetylene, n-pentylacetylene, pubchem13058, 1-heptyne, acmc-1aw2o
hept-1-yne
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement
P210-P243-P301+P330+P331-P315-P403+P233
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take precautionary measures against static discharge.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Get immediate medical advice/attention.
Store in a well-ventilated place.
Keep container tightly closed.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

TSCA : TSCA

SDS Documents