Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Heptyn-3-ol, 98%

Chemical Identifiers

Chemical Name or Material 1-Heptyn-3-ol
CAS 7383-19-9
Molecular Formula C7H12O
Formula Weight 112.17
MDL Number MFCD00041597
UN Number UN1987
Beilstein 1740804
Synonym n-Butyl ethynyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0533006

  SDS
alfa aesar™
L0533006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 112.17
Chemical Name or Material 1-Heptyn-3-ol
CAS 7383-19-9
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Synonym n-Butyl ethynyl carbinol
Health Hazard 2 GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.
Health Hazard 1 Warning
UN Number UN1987
MDL Number MFCD00041597
Boiling Point 161° to 163°C
Beilstein 1740804
Molecular Formula C7H12O
Odor Recognizable
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Quantity 5g
Flash Point 59°C (138°F)

Applications
1-Heptyn-3-ol, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, heavy metal powders.

Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.