Please login to your online account to display your discounted pricing

Alfa Aesar™ (1-Heptyl)triphenylphosphonium bromide, 98+%

$26.68 - $26.68

Chemical Identifiers

Chemical Name or Material (1-Heptyl)triphenylphosphonium bromide
CAS 13423-48-8
Molecular Formula C25H30BrP
Linear Formula CH3(CH2)6P(C6H5)3Br
Formula Weight 441.39
MDL Number MFCD00050249
Beilstein 3580843
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2542406

  SDS
alfa aesar™
B2542406
5g Each for $26.68

AAB2542414

  SDS
alfa aesar™
B2542414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material (1-Heptyl)triphenylphosphonium bromide
Odor Odorless
Melting Point 176° to 180°C
Quantity 5g
CAS 13423-48-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay ≥98%
Molecular Formula C25H30BrP
Linear Formula CH3(CH2)6P(C6H5)3Br
Formula Weight 441.39
MDL Number MFCD00050249
Beilstein 3580843
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.